Learn More
4'-Chloro-2'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals
Catalog Number p-4813088
Click to view available options
Quantity:
10 g
50 g
6921-66-0
C8H7ClO2
170.59
QCVSDCHNBNFJDQ-UHFFFAOYSA-N
1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
1051513
1-(4-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)O
6921-66-0
C8H7ClO2
170.59
QCVSDCHNBNFJDQ-UHFFFAOYSA-N
1-4-chloro-2-hydroxyphenyl ethanone, 4'-chloro-2'-hydroxyacetophenone, 4-chloro-2-hydroxyacetophenone, 1-4-chloro-2-hydroxyphenyl ethan-1-one, 2'-hydroxy-4'-chloroacetophenone, 1-4-chloro-2-hydroxy-phenyl ethanone, ethanone, 1-4-chloro-2-hydroxyphenyl, 4'-chloro-2'-hydroxy-acetophenone, ethanone,1-4-chloro-2-hydroxyphenyl
1051513
1-(4-chloro-2-hydroxyphenyl)ethanone
CC(=O)C1=C(C=C(C=C1)Cl)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 27°C to 30°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CLC6H3(OH)COCH3
Quantity 10 g
Solubility Information Solubility in water: insoluble
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-hydroxyacetophenone
compliance-icons
Product Identifier
  • 4'-Chloro-2'-hydroxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.