missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorophenylboronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 11966161
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
160591-91-3
C6H5BClFO2
174.37
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
4193595
(4-chloro-2-fluorophenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)F)(O)O
This item is not returnable. View return policy
160591-91-3
C6H5BClFO2
174.37
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
4193595
(4-chloro-2-fluorophenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)F)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 233.5°C to 235.5°C
Boiling Point 289.2°C
Flash Point 128.7°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 174.37
Percent Purity 95%
Chemical Name or Material 4-Chloro-2-fluorophenylboronic acid
compliance-icons
Product Identifier
  • 4-Chloro-2-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'