missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoro-5-nitrotoluene, 98%, Thermo Scientific™

Catalog Number p-7041365
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
18349-11-6
C7H5ClFNO2
189.57
MFCD00134231
SJDPAVRCQNFVDM-UHFFFAOYSA-N
87593
1-chloro-5-fluoro-4-methyl-2-nitrobenzene
CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O
This item is not returnable. View return policy
18349-11-6
C7H5ClFNO2
189.57
MFCD00134231
SJDPAVRCQNFVDM-UHFFFAOYSA-N
87593
1-chloro-5-fluoro-4-methyl-2-nitrobenzene
CC1=CC(=C(Cl)C=C1F)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 42°C to 45°C
Flash Point 113°C (235°F)
Quantity 5 g
Synonym 4-chloro-2-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-4-methyl-2-nitro-benzene, toluene, 4-chloro-2-fluoro-5-nitro, benzene,1-chloro-5-fluoro-4-methyl-2-nitro, benzene, 1-chloro-5-fluoro-4-methyl-2-nitro, acmc-1c7av, 4-chloro-2-fluoro-5-nitro-toluene, buttpark 146\15-31, 2-chloro-4-floro-5-methylnitrobenzene, 2-chloro-4-fluoro-5-methylnitrobenzene
Formula Weight 189.57
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoro-5-nitrotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'