missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-difluorobenzaldehyde, 98%, Thermo Scientific™

Catalog Number p-7055511
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
879093-02-4
C7H3ClF2O
176.55
MFCD07782051
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
18991142
4-chloro-2,5-difluorobenzaldehyde
FC1=CC(Cl)=C(F)C=C1C=O
This item is not returnable. View return policy
879093-02-4
C7H3ClF2O
176.55
MFCD07782051
XPZYVMBPIANKJG-UHFFFAOYSA-N
4-chloro-2,5-difluoro-benzaldehyde, benzaldehyde, 4-chloro-2,5-difluoro, 2,5-difluoro-4-chlorobenzaldehyde, pubchem1430, benzaldehyde,4-chloro-2,5-difluoro
18991142
4-chloro-2,5-difluorobenzaldehyde
FC1=CC(Cl)=C(F)C=C1C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 14516595
Formula Weight 176.55
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Chloro-2,5-difluorobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'