Få mere at vide
(4-Carboxybutyl-d4)triphenylphosphonium Bromide, TRC
Artikelnummer. 30349912
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30349912 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349912 Leverandør Toronto Research Chemicals Leverandørnr. TRCC177817250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4-Carboxybutyl-d4)triphenylphosphonium Bromide
CAS 42932-63-8
Molekylær formel C23 D4 H20 O2 P . Br
Mængde 250 mg
Synonym Phosphonium, (4-carboxybutyl-2,2,3,3-d4)triphenyl-, bromide (1:1) (ACI), Phosphonium, (4-carboxybutyl-2,2,3,3-d4)triphenyl-, bromide (9CI), Triphenylphosphonium 5-bromovalerate-3,3,4,4-d4, (4-Carboxybutyl-d4)triphenylphosphonium Bromide, (4-Carboxybutyl-2,2,3,3-d4)triphenylphosphonium Bromide, (4-Carboxybutyl-2,2,3,3-D4)triphenylphosphonium Bromide, (4-Carboxybutyl)triphenylphosphonium-D4 Bromide
InChI formel InChI=1S/C23H23O2P.BrH/c24-23(25)18-10-11-19-26(20-12-4-1-5-13-20,21-14-6-2-7-15-21)22-16-8-3-9-17-22;/h1-9,12-17H,10-11,18-19H2;1H/i10D2,11D2;
SMIL [Br-].[2H]C([2H])(CC(=O)O)C([2H])([2H])C[P+](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC navn (4-carboxy-2,2,3,3-tetradeuteriobutyl)-triphenylphosphanium;bromide
Molekylvægt (g/mol) 447.3379
Formel vægt 446.0948