missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 11472338
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
619-66-9
C8H6O3
150.133
MFCD00006951
GOUHYARYYWKXHS-UHFFFAOYSA-N
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
12088
4-formylbenzoic acid
C1=CC(=CC=C1C=O)C(=O)O
This item is not returnable. View return policy
619-66-9
C8H6O3
150.133
MFCD00006951
GOUHYARYYWKXHS-UHFFFAOYSA-N
4-carboxybenzaldehyde, benzoic acid, 4-formyl, terephthalaldehydic acid, p-formylbenzoic acid, p-carboxybenzaldehyde, terephthaldehydic acid, 4-carboxybenzaladehyde, 4-formyl-benzoic acid, 4-formylbenzoicacid, unii-ues4qrk36e
12088
4-formylbenzoic acid
C1=CC(=CC=C1C=O)C(=O)O

4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxybenzaldehyde is an important intermediate in the synthesis of terepthalic acid. It has been used as reagent during esterification of 2,2,6,6-tetramethyl-4-oxopiperidinyl-1-oxyl to yield 4-hydroxy-2,2,6,6-tetramethylpiperidine-1-oxyl 4-formylbenzoate.

Solubility
Soluble in water, methanol, DMSO, ether, and chloroform.

Notes
Store away from air and strong bases. Air Sensitive. Incompatible with bases and air.
TRUSTED_SUSTAINABILITY
Melting Point ∼245°C
Odor Odorless
Quantity 50 g
Beilstein 471734
Sensitivity Air Sensitive
Solubility Information Soluble in water,methanol,DMSO,ether,and chloroform.
Formula Weight 150.13
Percent Purity 98%
Chemical Name or Material 4-Carboxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'