Få mere at vide
4-(Butylamino)benzoic Acid, Mikromol™
Artikelnummer. 17902036
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17902036 25 mg
17961676 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17902036 Leverandør Mikromol Leverandørnr. MM0504.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-(Butylamino)benzoic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Butylamino)benzoic Acid
CAS 4740-24-3
Analyt- eller komponentnavne 4-(Butylamino)benzoic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N O2
Certificeringer/compliance ISO 17025
Synonym Tetracaine Hydrochloride Imp. B (EP), Tetracaine Imp. B (EP), Tetracaine USP Related Compound B, Tetracaine USP RC B, 4-(Butylamino)benzoic Acid, Tetracaine Related Compound B, Tetracaine Hydrochloride Impurity B, Tetracaine Impurity B
InChI formel InChI=1S/C11H15NO2/c1-2-3-8-12-10-6-4-9(5-7-10)11(13)14/h4-7,12H,2-3,8H2,1H3,(H,13,14)
SMIL CCCCNc1ccc(cc1)C(=O)O
IUPAC navn 4-(butylamino)benzoic acid
Molekylvægt (g/mol) 193.24
Mængde 25 mg
Formel vægt 193.1103 g/mol