Få mere at vide
4-Bromotoluene-2,3,5,6-D4, TRC
Artikelnummer. 30339355
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30339355 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339355 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68845910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromotoluene-2,3,5,6-D4
CAS 112484-85-2
Molekylær formel C7 D4 H3 Br
Mængde 10 mg
Synonym Benzene-1,2,4,5-d4, 3-bromo-6-methyl- (9CI, ACI), 3-Bromo-6-methylbenzene-1,2,4,5-d4 (ACI), Toluene-2,3,5,6-d4, p-bromo- (6CI), 1-Bromotetradeutero-4-methylbenzene, 4-Bromotoluene-d4, 4-Bromotoluene-2,3,5,6-d4, 4-Bromotoluene-2,3,5,6-D4
InChI formel InChI=1S/C7H7Br/c1-6-2-4-7(8)5-3-6/h2-5H,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(Br)c([2H])c([2H])c1C
IUPAC navn 1-bromo-2,3,5,6-tetradeuterio-4-methylbenzene
Molekylvægt (g/mol) 175.0591
Formel vægt 173.9982