missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromothiobenzamide, 97%

Brand:  Alfa Aesar™ H55956.06

Additional Details : CAS Number : 26197-93-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15472358

  • 1215.65 DKK / 5g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

26197-93-3
216.096
AIPANIYQEBQYGC-UHFFFAOYSA-N
1549289
C1=CC(=CC=C1C(=S)N)Br
C7H6BrNS
MFCD04114360
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
4-bromobenzenecarbothioamide

Specifications

4-Bromothiobenzamide
139°C to 143°C
26197-93-3
MFCD04114360
AIPANIYQEBQYGC-UHFFFAOYSA-N
4-bromobenzenecarbothioamide
1549289
Solid
Brown to White
5g
C7H6BrNS
4-bromothiobenzamide, 4-bromo-thiobenzamide, p-bromothiobenzamide, 4-bromobenzothioamide, benzenecarbothioamide, 4-bromo, 4-bromobenzene-1-carbothioamide, amino 4-bromophenyl methane-1-thione, acmc-1cehr, 4-bromanylbenzenecarbothioamide
C1=CC(=CC=C1C(=S)N)Br
216.096
216.1
97%
Videos
SDS Documents