Learn More
4-Bromopyrene, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromopyrene |
| CAS | 1732-26-9 |
| Molecular Formula | C16H9Br |
| Quantity | 25 mg |
| Synonym | Pyrene, 4-bromo-, 4-Bromopyrene |
| InChI Formula | InChI=1S/C16H9Br/c17-14-9-12-5-1-3-10-7-8-11-4-2-6-13(14)16(11)15(10)12/h1-9H |
| SMILES | Brc1cc2cccc3ccc4cccc1c4c23 |
| IUPAC Name | 4-bromopyrene |
| Molecular Weight (g/mol) | 281.15 |
| Formula Weight | 279.9888 |