Learn More
4'-Bromopropiophenone, 97%
Product Code. 11496907
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496907 10 g 10g
11334769 25 g 25g
11406917 50 g 50g
11416917 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 11496907 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A14349.09
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
CAS: 10342-83-3 | C9H9BrO | 213.07 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 10342-83-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00000106
InChI Key UOMOSYFPKGQIKI-UHFFFAOYSA-N
Synonym 4'-bromopropiophenone, 1-4-bromophenyl propan-1-one, p-bromopropiophenone, 4-bromopropiophenone, propiophenone, 4'-bromo, 4-bromophenyl ethyl ketone, 1-propanone, 1-4-bromophenyl, 1-4-bromophenyl-1-propanone, aurora 22532
PubChem CID 66312
IUPAC Name 1-(4-bromophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(Br)C=C1
Tekniske data
Melting Point 45°C to 47°C
Boiling Point 129°C to 131°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
Beilstein 1210311
Formula Weight 213.08
Percent Purity 97%
Chemical Name or Material 4'-Bromopropiophenone