missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromophthalimide, ≥97%

Brand:  Alfa Aesar™ H64136.03

Additional Details : CAS Number : 6941-75-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471199

  • 196.89 DKK / 1g


In Stock 2
Add to basket

Description

Description

Applications
It is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Soluble in N,N-DMF almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

6941-75-9
226.029
GNYICZVGHULCHE-UHFFFAOYSA-N
236018
C1=CC2=C(C=C1Br)C(=O)NC2=O
C8H4BrNO2
MFCD00466049
4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
5-bromoisoindole-1,3-dione

Specifications

4-Bromophthalimide
230°C to 235°C
6941-75-9
MFCD00466049
4-bromophthalimide, 5-bromoisoindoline-1,3-dione, 5-bromo-isoindole-1,3-dione, 5-bromo-2,3-dihydro-1h-isoindole-1,3-dione, 4-bromo phthalimide, 5-bromo-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, 5-bromo, 5-bromo-2h-isoindole-1,3-dione, 5-bromophthalimide, acmc-209o8b
GNYICZVGHULCHE-UHFFFAOYSA-N
5-bromoisoindole-1,3-dione
236018
Solid
White
1g
C8H4BrNO2
131235
Soluble in N,N-DMF almost transparency.
C1=CC2=C(C=C1Br)C(=O)NC2=O
226.029
226.03
≥97%
Videos
SDS Documents