Få mere at vide
4-Bromophenol-2,3,5,6-d4, TRC
Artikelnummer. 30331196
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30331196 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331196 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68640750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromophenol-2,3,5,6-d4
CAS 152404-44-9
Molekylær formel C6 2H4 H Br O
Mængde 50 mg
Synonym Phen-2,3,5,6-d4-ol, 4-bromo-, 4-Bromophen-2,3,5,6-d4-ol, 4-Bromophenol D4, 4-Bromophenol (ring-D4), 4-Bromo-2,3,5,6-tetradeuteriophenol
InChI formel InChI=1S/C6H5BrO/c7-5-1-3-6(8)4-2-5/h1-4,8H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(Br)c([2H])c([2H])c1O
IUPAC navn 4-bromo-2,3,5,6-tetradeuteriophenol
Molekylvægt (g/mol) 177.03
Formel vægt 175.9775