Få mere at vide
4-Bromonitrobenzene-d4, CDN
Artikelnummer. 30313370
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313370 1 g -
30309593 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313370 Leverandør CDN Leverandørnr. CDND50231G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Bromo-4-nitrobenze-d4
CAS 350820-19-8
Molekylær formel C6 2H4 Br N O2
Mængde 1 g
InChI formel InChI=1 S/C6H4BrNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Br)[N+](=O)[O-]
IUPAC navn 1-bromo-2,3,5,6-tetradeuterio-4-nitrobenzene
Molekylvægt (g/mol) 206.03
Formel vægt 204.9676 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity