missing translation for 'onlineSavingsMsg'
Learn More

4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number p-7040151
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1029439-49-3
C13H17BBrFO2
314.989
MFCD18434471
QOCYLABQSVWXGE-UHFFFAOYSA-N
2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester
53217331
2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F
This item is not returnable. View return policy
1029439-49-3
C13H17BBrFO2
314.989
MFCD18434471
QOCYLABQSVWXGE-UHFFFAOYSA-N
2-4-bromomethyl-2-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-bromomethyl-2-fluorophenylboronic acid pinacol ester, 4-bromomethyl-2-fluorobenzeneboronic acid pinacol ester, 4-bromomethyl-2-fluorophenyl boronic acid pinacol ester
53217331
2-[4-(bromomethyl)-2-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)CBr)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 64°C
Quantity 250 mg
Formula Weight 314.99
Percent Purity 96%
Chemical Name or Material 4-Bromomethyl-2-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'