Få mere at vide
4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane-d5, TRC
Artikelnummer. 30329715
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30329715 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329715 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68534210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane-d5
CAS 1346603-73-3
Molekylær formel C6H6D5BrO2
Mængde 10 mg
Synonym 4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane-d5, DL-Isopropylideneglycerol-1-bromohydrin-d5
InChI formel InChI=1S/C6H11BrO2/c1-6(2)8-4-5(3-7)9-6/h5H,3-4H2,1-2H3/i3D2,4D2,5D
SMIL [2H]C([2H])(Br)C1([2H])OC(C)(C)OC1([2H])[2H]
IUPAC navn 4-[bromo(dideuterio)methyl]-4,5,5-trideuterio-2,2-dimethyl-1,3-dioxolane
Molekylvægt (g/mol) 200.09
Formel vægt 199.0256