Få mere at vide
4-Bromobenzyl-d6 Bromide, CDN
Artikelnummer. 30328201
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328201 0.5 g 0.5g
30309304 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328201 Leverandør CDN Leverandørnr. CDND75090.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromobenzyl-d6 Bromide
CAS 229948-68-9
Molekylær formel C7 2H6 Br2
Mængde 0.5 g
Synonym Benzene-1,2,4,5-d4, 3-bromo-6-(bromomethyl-d2)- (9 CI), 4-Bromobenzyl-d6 Bromide, 1-Bromo-4-[bromo(dideuterio)methyl]-2,3,5,6-tetradeuteriobenzene
InChI formel InChI=1 S/C7H6Br2/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D,5D2
SMIL [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Br)C([2 H])([2 H])Br
IUPAC navn 1-bromo-4-[bromo(dideuterio)methyl]-2,3,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 255.968
Formel vægt 253.921 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity