missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7045651
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
589-17-3
C7H6BrCl
205.48
MFCD00040867
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
68528
1-bromo-4-(chloromethyl)benzene
ClCC1=CC=C(Br)C=C1
This item is not returnable. View return policy
589-17-3
C7H6BrCl
205.48
MFCD00040867
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
68528
1-bromo-4-(chloromethyl)benzene
ClCC1=CC=C(Br)C=C1

4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 41°C
Boiling Point 106°C to 107°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 6123220
Merck Index 14,1410
Solubility Information Insoluble in water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 4-Bromobenzyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'