Få mere at vide
4-Bromobenzyl-2,3,5,6-d4 Bromide, CDN
Artikelnummer. 30324140
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324140 0.25 g
30312601 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324140 Leverandør CDN Leverandørnr. CDND75400.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromobenzyl Bromide-2,3,5,6-D4
CAS 2708280-84-4
Molekylær formel C7 D4 H2 Br2
Mængde 0.25 g
Synonym 3-Bromo-6-bromomethylbenzene-1,2,4,5-d4, 4-Bromobenzyl-d4 Bromide, 4-Bromobenzyl-2,3,5,6-d4 Bromide, 1-Bromo-4-(bromomethyl)-2,3,5,6-tetradeuteriobenzene, 1-Bromomethyl-4-bromobenzene-2,3,5,6-d4, 4-Bromobenzyl Bromide-2,3,5,6-D4
InChI formel InChI=1 S/C7H6Br2/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(CBr)c([2 H])c([2 H])c1Br
IUPAC navn 1-bromo-4-(bromomethyl)-2,3,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 253.9552
Formel vægt 251.9087 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity