Få mere at vide
4-Bromobenzoic-d4 Acid, CDN
Artikelnummer. 30315856
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315856 0.5 g 0.5g
30320938 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315856 Leverandør CDN Leverandørnr. CDND62930.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromobenzoic Acid-D4
CAS 787624-24-2
Molekylær formel C7 D4 H Br O2
Mængde 0.5 g
Synonym Benzoic-2,3,5,6-d4 acid, 4-bromo- (9 CI), 4-Bromobenzoic-2,3,5,6-d4 acid, 4-Bromobenzoic-d4 Acid, 4-Bromobenzoic Acid-d4, 4-Bromobenzoic Acid-D4, p-Bromobenzoic Acid-d4
InChI formel InChI=1 S/C7H5BrO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1Br
IUPAC navn 4-bromo-2,3,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 205.0421
Formel vægt 203.9724 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity