missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromobenzaldehyde dimethyl acetal, 98%

Brand:  Alfa Aesar™ H55283.AE

Additional Details : CAS Number : 24856-58-4 Weight : 0.00010kg

Quantity 100mL

 View more versions of this product

Product Code. 15401498

  • 2261.95 DKK / 100mL


Estimated Shipment date
from Supplier 22-01-2019
Add to basket

Description

Description

Applications
4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

24856-58-4
231.089
TWFNGPDYMFEHOB-UHFFFAOYSA-N
688342
COC(C1=CC=C(C=C1)Br)OC
C9H11BrO2
MFCD01320499
4-bromobenzaldehyde dimethyl acetal, 1-bromo-4-dimethoxymethyl benzene, acmc-20ammy, pubchem5380, 4-bromophenyl dimethoxymethane, 1-bromo-4-dimethoxymethyl benzene, 1-bromo-4-dimethoxymethyl-benzene, 4-bromobenzaldehyde dimethyl ketal, 4-bromo-benzaldehyde dimethyl ketal, 4-bromobenzaldehyde dimethyl acetal
1-bromo-4-(dimethoxymethyl)benzene

Specifications

4-Bromobenzaldehyde dimethyl acetal
1.5320
100mL
C9H11BrO2
4-bromobenzaldehyde dimethyl acetal, 1-bromo-4-dimethoxymethyl benzene, acmc-20ammy, pubchem5380, 4-bromophenyl dimethoxymethane, 1-bromo-4-dimethoxymethyl benzene, 1-bromo-4-dimethoxymethyl-benzene, 4-bromobenzaldehyde dimethyl ketal, 4-bromo-benzaldehyde dimethyl ketal, 4-bromobenzaldehyde dimethyl acetal
COC(C1=CC=C(C=C1)Br)OC
231.089
231.09
98%
1.383g/mL
254°C
24856-58-4
MFCD01320499
TWFNGPDYMFEHOB-UHFFFAOYSA-N
1-bromo-4-(dimethoxymethyl)benzene
688342
Liquid
Videos
SDS