missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde dimethyl acetal, 97%, Thermo Scientific™

Catalog Number 15491488
Click to view available options
Quantity:
25 mL
This item is not returnable. View return policy
24856-58-4
C9H11BrO2
231.089
MFCD01320499
TWFNGPDYMFEHOB-UHFFFAOYSA-N
4-bromobenzaldehyde dimethyl acetal, 1-bromo-4-dimethoxymethyl benzene, acmc-20ammy, pubchem5380, 4-bromophenyl dimethoxymethane, 1-bromo-4-dimethoxymethyl-benzene, 4-bromobenzaldehyde dimethyl ketal, 4-bromo-benzaldehyde dimethyl ketal
688342
1-bromo-4-(dimethoxymethyl)benzene
COC(C1=CC=C(C=C1)Br)OC
This item is not returnable. View return policy
24856-58-4
C9H11BrO2
231.089
MFCD01320499
TWFNGPDYMFEHOB-UHFFFAOYSA-N
4-bromobenzaldehyde dimethyl acetal, 1-bromo-4-dimethoxymethyl benzene, acmc-20ammy, pubchem5380, 4-bromophenyl dimethoxymethane, 1-bromo-4-dimethoxymethyl-benzene, 4-bromobenzaldehyde dimethyl ketal, 4-bromo-benzaldehyde dimethyl ketal
688342
1-bromo-4-(dimethoxymethyl)benzene
COC(C1=CC=C(C=C1)Br)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.383g/mL
Boiling Point 254°C
Refractive Index 1.5320
Quantity 25 mL
Formula Weight 231.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'