Få mere at vide
4’-Bromoacetophenone-d4, TRC
Artikelnummer. 30336652
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30336652 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336652 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67906225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4'-Bromoacetophenone-d4
CAS 343942-02-9
Molekylær formel C8 2H4 H3 Br O
Mængde 25 mg
Synonym 1-(4-Bromophenyl)ethanone-d4, p-Bromoacetophenone-d4, 1-(4-Bromophenyl)ethanone-d4, 1-(p-Bromophenyl)ethanone-d4, 1-Acetyl-4-bromobenzene-d4, 1-Bromo-4-acetylbenzene-d4, 4-Acetyl-1-bromobenzene-d4, 4-Acetylbromobenzene-d4, 4-Acetylphenyl-d4 Bromide, 4-Bromobenzene-d4 Methyl Ketone, 4-Bromohypnone-d4, 4-Bromophenyl-d4 Methyl Ketone
InChI formel InChI=1S/C8H7BrO/c1-6(10)7-2-4-8(9)5-3-7/h2-5H,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(C(=O)C)c([2H])c([2H])c1Br
IUPAC navn 1-(4-bromo-2,3,5,6-tetradeuteriophenyl)ethanone
Molekylvægt (g/mol) 203.069
Formel vægt 201.993