Learn More
Alfa Aesar™ 4-Bromo-N-(4-bromo-3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63068.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15408116
  • 9607.87 DKK / 5g
Estimated Shipment date
from Supplier 31-01-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(4-bromo-3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
477.07
MFCD26143353
Specifications
4-Bromo-N-(4-bromo-3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
5g
477.07
Solid
97%
MFCD26143353