Learn More
Alfa Aesar™ 4-Bromo-N-(3-fluoro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H63275.06
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
4-Bromo-N-(3-fluoro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide | |
412.2 | |
MFCD28098714 |
Specifications
4-Bromo-N-(3-fluoro-2-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide | |
97% | |
412.2 |
Solid | |
5g | |
MFCD28098714 |