Learn More
Alfa Aesar™ 4-Bromo-N-(3-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H62568.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15407586
  • 2403.83 DKK / 1g
Estimated Shipment date
from Supplier 01-08-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(3-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143330
Specifications
4-Bromo-N-(3-chlorophenyl)-3-(trifluoromethyl)benzenesulfonamide
Solid
97%
414.63
1g
MFCD26143330