missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65498.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469686

  • 2009.39 DKK / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide
390.22
MFCD27975769

Specifications

4-Bromo-N-(2,5-dimethoxyphenyl)-2-fluorobenzenesulfonamide
1g
390.22
Solid
97%
MFCD27975769
Videos
SDS