missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine, 99%, Thermo Scientific™

Catalog Number p-74166
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14091-15-7
C9H10BrNO2
244.09
MFCD00002599
PEMUHKUIQHFMTH-UHFFFAOYSA-N
2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
85681
2-amino-3-(4-bromophenyl)propanoic acid
NC(CC1=CC=C(Br)C=C1)C(O)=O
This item is not returnable. View return policy
14091-15-7
C9H10BrNO2
244.09
MFCD00002599
PEMUHKUIQHFMTH-UHFFFAOYSA-N
2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
85681
2-amino-3-(4-bromophenyl)propanoic acid
NC(CC1=CC=C(Br)C=C1)C(O)=O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 262.0°C to 263.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4CH2CH(NH2)CO2H
Quantity 250 mg
Formula Weight 244.09
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Bromo-DL-phenylalanine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'