Få mere at vide
4-Bromo-a-phenylbenzenemethanol-d5, TRC
Artikelnummer. 30347869
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30347869 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347869 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6802721G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromo-alpha-phenylbenzenemethanol-d5
CAS 213678-51-4
Molekylær formel C13H6D5BrO
Mængde 1 g
Synonym 4-Bromobenzhydrol-d5, 4-Bromobenzohydrol-d5, (4-Bromophenyl)phenylmethanol-d5, (±)-(4-Bromophenyl)phenylmethanol-d5, 4-(1-Hydroxybenzyl)bromobenzene-d5, 4-Bromobenzhydrol-d5, NSC 89817-d5, p-Bromobenzhydrol-d5, alpha-(4-Bromophenyl)benzenemethanol-d5
InChI formel InChI=1S/C13H11BrO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13,15H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(O)c2ccc(Br)cc2)c([2H])c1[2H]
IUPAC navn (4-bromophenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanol
Molekylvægt (g/mol) 268.16
Formel vægt 267.0307