Learn More
4-Bromo-3-methylaniline, 97%, Thermo Scientific™
Catalog Number p-75714
Click to view available options
Quantity:
10 g
50 g
250 g
6933-10-4
C7H8BrN
186.05
MFCD00007828
6933-10-4
C7H8BrN
186.05
MFCD00007828
CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 83.0°C
Color Brown to White
Boiling Point 240.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(CH3)NH2
Quantity 10 g
Beilstein 12, 873
Synonym 4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
Solubility Information Solubility in water: insoluble
InChI Key MMEGELSFOYDPQW-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)N)Br
IUPAC Name 4-bromo-3-methylaniline
PubChem CID 23359
Formula Weight 186.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-3-methylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.