Learn More
4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is used as a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 181°C to 185°C |
Quantity | 250 mg |
Beilstein | 9620931 |
Formula Weight | 237 |
Percent Purity | 96% |
Chemical Name or Material | 4-Bromo-3,5-difluorobenzoic acid |