missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluorobenzoic acid, 96%, Thermo Scientific Chemicals

Catalog Number 15406358
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
651027-00-8
C7H3BrF2O2
237.00
MFCD18917156
NLLRAEQVOZOSBB-UHFFFAOYSA-N
benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
11424897
4-bromo-3,5-difluorobenzoic acid
OC(=O)C1=CC(F)=C(Br)C(F)=C1
This item is not returnable. View return policy
651027-00-8
C7H3BrF2O2
237.00
MFCD18917156
NLLRAEQVOZOSBB-UHFFFAOYSA-N
benzoic acid, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluorobenzoicacid, 3,5-difluoro-4-bromobenzoic acid
11424897
4-bromo-3,5-difluorobenzoic acid
OC(=O)C1=CC(F)=C(Br)C(F)=C1

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 181°C to 185°C
Quantity 250 mg
Beilstein 9620931
Formula Weight 237
Percent Purity 96%
Chemical Name or Material 4-Bromo-3,5-difluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'