Få mere at vide
4-Bromo-2-(trifluoromethyl)benzonitrile, TRC
Artikelnummer. 30331856
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30331856 250 mg
30343439 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30331856 Leverandør Toronto Research Chemicals Leverandørnr. TRCB695508250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromo-2-(trifluoromethyl)benzonitrile
CAS 191165-13-6
Molekylær formel C8H3BrF3N
Mængde 250 mg
InChI formel InChI=1S/C8H3BrF3N/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3H
SMIL FC(F)(F)c1cc(Br)ccc1C#N
IUPAC navn 4-bromo-2-(trifluoromethyl)benzonitrile
Molekylvægt (g/mol) 250.02
Formel vægt 248.9401