missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-(methylthio)pyrimidine, 97%

Brand:  Alfa Aesar™ H64124.03

Additional Details : CAS Number : 959236-97-6 Weight : 0.00100kg Transport : UN number : 1759 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15471179

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance
View alternative products »


Specifications

Chemical Identifiers

959236-97-6
205.073
TZSQAGYDWPPKPE-UHFFFAOYSA-N
45787531
CSC1=NC=CC(=N1)Br
C5H5BrN2S
MFCD09878932
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine, 2-methylthio-4-bromopyrimidine
4-bromo-2-methylsulfanylpyrimidine

Specifications

4-Bromo-2-(methylthio)pyrimidine
30°C to 40°C
959236-97-6
MFCD09878932
21913366
TZSQAGYDWPPKPE-UHFFFAOYSA-N
4-bromo-2-methylsulfanylpyrimidine
45787531
Low Melting Solid
Brown to Yellow
1g
C5H5BrN2S
UN1759
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine, 2-methylthio-4-bromopyrimidine
CSC1=NC=CC(=N1)Br
205.073
205.07
97%
Videos
SDS