Learn More
4-Bromo-2-fluorobenzylamine hydrochloride, 98%, Thermo Scientific™
Catalog Number 11497567
Click to view available options
Quantity:
5 g
25 g
100 g
147181-08-6
C7H8BrClFN
240.5
MFCD00143266
RTQOFJWBJNVJEO-UHFFFAOYSA-N
4-bromo-2-fluorobenzylamine hydrochloride, 4-bromo-2-fluorophenyl methanamine hydrochloride, 4-bromo-2-fluorobenzylamine hcl, 1-4-bromo-2-fluorophenyl methanamine hydrochloride, benzenemethanamine, 4-bromo-2-fluoro-, hydrochloride, 4-bromo-2-fluorobenzylamine hydrochlorid, pubchem3774, pubchem12660, acmc-1by0k, 4-bromo-2-fluoro-benzylamine hydrochloride
18468197
(4-bromo-2-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1Br)F)CN.Cl
147181-08-6
C7H8BrClFN
240.5
MFCD00143266
RTQOFJWBJNVJEO-UHFFFAOYSA-N
4-bromo-2-fluorobenzylamine hydrochloride, 4-bromo-2-fluorophenyl methanamine hydrochloride, 4-bromo-2-fluorobenzylamine hcl, 1-4-bromo-2-fluorophenyl methanamine hydrochloride, benzenemethanamine, 4-bromo-2-fluoro-, hydrochloride, 4-bromo-2-fluorobenzylamine hydrochlorid, pubchem3774, pubchem12660, acmc-1by0k, 4-bromo-2-fluoro-benzylamine hydrochloride
18468197
(4-bromo-2-fluorophenyl)methanamine;hydrochloride
C1=CC(=C(C=C1Br)F)CN.Cl
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 254°C to 257°C
Quantity 5 g
Beilstein 6938847
Formula Weight 240.5
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzylamine hydrochloride