missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluorobenzonitrile, 99%

Brand:  Alfa Aesar™ A13086.03

Additional Details : CAS Number : 105942-08-3 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11494404

  • 167.00 DKK / 1g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

105942-08-3
200.01
HGXWRDPQFZKOLZ-UHFFFAOYSA-N
736029
C1=CC(=C(C=C1Br)F)C#N
C7H3BrFN
MFCD00143264
2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
4-bromo-2-fluorobenzonitrile

Specifications

4-Bromo-2-fluorobenzonitrile
69°C to 72°C
105942-08-3
MFCD00143264
4231143
Insoluble in water.
C1=CC(=C(C=C1Br)F)C#N
200.01
200.01
238°C to 239°C
1g
C7H3BrFN
UN3439
2-fluoro-4-bromobenzonitrile, 4-bromo-2-fluoro-benzonitrile, benzonitrile, 4-bromo-2-fluoro, 4-bromo-2-fluorobenzenecarbonitrile, 4-cyano-3-fluorobromobenzene, pubchem3221, 2-floro-4-bromobenzonitrile, 4-bromo-2-fluorobenzontrile, 4-bromo-6-fluorobenzonitrile, ksc490q4t
HGXWRDPQFZKOLZ-UHFFFAOYSA-N
4-bromo-2-fluorobenzonitrile
736029
99%
Videos
SDS Documents