Learn More
Alfa Aesar™ 4-Bromo-2-fluoro-N-(4-fluorobenzyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64658.03
Additional Details : CAS Number : 1350926-53-2 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469056
  • 1908.81 DKK / 1g
Estimated Shipment date
from Supplier 20-02-2019
Add to basket
Specifications
Chemical Identifiers
1350926-53-2
362.189
HXFUECGMSBHDHG-UHFFFAOYSA-N
97455681
C1=CC(=CC=C1CNS(=O)(=O)C2=C(C=C(C=C2)Br)F)F
C13H10BrF2NO2S
MFCD27975690
n-4-flurobenzyl-4-bromo-2-fluoro-benzenesulfonamide, 4-bromo-2-fluoro-n-4-fluorophenyl methyl benzenesulfonamide
4-bromo-2-fluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
Specifications
4-Bromo-2-fluoro-N-(4-fluorobenzyl)benzenesulfonamide
1350926-53-2
MFCD27975690
HXFUECGMSBHDHG-UHFFFAOYSA-N
4-bromo-2-fluoro-N-[(4-fluorophenyl)methyl]benzenesulfonamide
97455681
Solid
1g
C13H10BrF2NO2S
n-4-flurobenzyl-4-bromo-2-fluoro-benzenesulfonamide, 4-bromo-2-fluoro-n-4-fluorophenyl methyl benzenesulfonamide
C1=CC(=CC=C1CNS(=O)(=O)C2=C(C=C(C=C2)Br)F)F
362.189
362.19
97%