Få mere at vide
4-bromo-2-chloro-N-methylaniline, TRC
Artikelnummer. 30180244
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180244 100 mg
30179855 25 mg
30180210 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180244 Leverandør Toronto Research Chemicals Leverandørnr. TRCB593840100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 944131-95-7
Molekylær formel C7 H7 Br Cl N
Molekylvægt (g/mol) 220.494
IUPAC navn 4-bromo-2-chloro-N-methylaniline
SMIL CNc1ccc(Br)cc1Cl
Tekniske data
Kemisk navn eller materiale 4-bromo-2-chloro-N-methylaniline
Mængde 100 mg
InChI formel InChI=1S/C7H7BrClN/c1-10-7-3-2-5(8)4-6(7)9/h2-4,10H,1H3
Formel vægt 218.945