missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-(4-morpholinyl)benzonitrile, 98%

Brand:  Alfa Aesar™ H61897.03

Additional Details : CAS Number : 1260762-06-8 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15438488

  • 2064.00 DKK / 1g

Estimated Shipment
29-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1260762-06-8
267.126
4-bromo-2-morpholin-4-yl benzonitrile, 4-bromo-2-morpholinobenzonitrile, acmc-209b1e
4-bromo-2-morpholin-4-ylbenzonitrile
C11H11BrN2O
SEDCSLWVDKMJCF-UHFFFAOYSA-N
70699567
C1COCCN1C2=C(C=CC(=C2)Br)C#N

Specifications

4-Bromo-2-(4-morpholinyl)benzonitrile
1260762-06-8
UN3439
SEDCSLWVDKMJCF-UHFFFAOYSA-N
4-bromo-2-morpholin-4-ylbenzonitrile
70699567
Solid
1g
C11H11BrN2O
4-bromo-2-morpholin-4-yl benzonitrile, 4-bromo-2-morpholinobenzonitrile, acmc-209b1e
C1COCCN1C2=C(C=CC(=C2)Br)C#N
267.126
267.13
98%
Videos
SDS Documents