missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7048044
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
122033-54-9
C7BrClF4O
291.424
MFCD03001173
XVYIQLOULDERAO-UHFFFAOYSA-N
4-bromo-2,3,5,6-tetrafluorobenzoylchloride, benzoyl chloride,4-bromo-2,3,5,6-tetrafluoro, acmc-20amnz, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoyl chloride
2783245
4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)Cl
This item is not returnable. View return policy
122033-54-9
C7BrClF4O
291.424
MFCD03001173
XVYIQLOULDERAO-UHFFFAOYSA-N
4-bromo-2,3,5,6-tetrafluorobenzoylchloride, benzoyl chloride,4-bromo-2,3,5,6-tetrafluoro, acmc-20amnz, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoyl chloride
2783245
4-bromo-2,3,5,6-tetrafluorobenzoyl chloride
C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 95°C to 100°C
Refractive Index 1.517
Quantity 1 g
UN Number UN3265
Beilstein 7295811
Sensitivity Moisture sensitive
Formula Weight 291.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'