missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Bromo-2-(2-nitro-1-propenyl)thiophene, 97%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 88°C to 90°C |
Quantity | 1 g |
Formula Weight | 248.09 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-2-(2-nitro-1-propenyl)thiophene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'