Få mere at vide
4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene, Mikromol
Artikelnummer. 30459083
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459083 4 x 25 mg
17982576 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30459083 Leverandør Mikromol Leverandørnr. MM3611.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene
CAS 461432-23-5
Analyt- eller komponentnavne 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 Br Cl O
Certificeringer/compliance ISO 17025
Synonym 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene, 4-bromo-1-chloro-2-[(4-ethoxyphenyl)methyl]benzene
InChI formel InChI=1S/C15H14BrClO/c1-2-18-14-6-3-11(4-7-14)9-12-10-13(16)5-8-15(12)17/h3-8,10H,2,9H2,1H3
Løsningstype Neat
SMIL CCOc1ccc(Cc2cc(Br)ccc2Cl)cc1
IUPAC navn 4-bromo-1-chloro-2-[(4-ethoxyphenyl)methyl]benzene
Molekylvægt (g/mol) 325.63
Mængde 4 x 25 mg
Formel vægt 323.9917 g/mol