Få mere at vide
4-Bromo-1-butanol-1,1,2,2,3,3,4,4-d8, CDN
Artikelnummer. 30326126
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326126 0.25 g -
30316984 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326126 Leverandør CDN Leverandørnr. CDND57040.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Bromo-1-butanol-D8
CAS 136091-68-4
Molekylær formel C4 D8 H Br O
Mængde 0.25 g
Synonym 1-Butan-1,1,2,2,3,3,4,4-d8-ol, 4-bromo- (9 CI), 4-Bromo-1-butanol-1,1,2,2,3,3,4,4-d8, 4-Bromo-1-butanol-D8, 1-Bromo-4-hydroxybutane-d8
InChI formel InChI=1 S/C4H9BrO/c5-3-1-2-4-6/h6H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br
IUPAC navn 4-bromo-1,1,2,2,3,3,4,4-octadeuteriobutan-1-ol
Molekylvægt (g/mol) 161.067
Formel vægt 160.0339 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity