Learn More
Alfa Aesar™ 4-(Boc-amino)indole-3-carboxylic acid, 97%
Brand:  Alfa Aesar™ H63752.MD
Additional Details : CAS Number : 885266-66-0 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15400609
  • 3004.88 DKK / 250mg
Estimated Shipment date
from Supplier 16-01-2019
Add to basket
Specifications
Chemical Identifiers
885266-66-0
276.292
QBNHICJNKZTOJB-UHFFFAOYSA-N
24720941
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C(=O)O
C14H16N2O4
MFCD04973970
4-tert-butoxycarbonyl amino-1h-indole-3-carboxylic acid, 4-tert-butoxycarbonylamino-1h-indole-3-carboxylic acid
4-[(2-methylpropan-2-yl)oxycarbonylamino]-1H-indole-3-carboxylic acid
Specifications
4-(Boc-amino)indole-3-carboxylic acid
250mg
C14H16N2O4
4-tert-butoxycarbonyl amino-1h-indole-3-carboxylic acid, 4-tert-butoxycarbonylamino-1h-indole-3-carboxylic acid
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C(=O)O
276.292
276.29
97%
White
885266-66-0
MFCD04973970
QBNHICJNKZTOJB-UHFFFAOYSA-N
4-[(2-methylpropan-2-yl)oxycarbonylamino]-1H-indole-3-carboxylic acid
24720941
Solid