Få mere at vide
4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8, TRC
Artikelnummer. 30346515
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30346515 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346515 Leverandør Toronto Research Chemicals Leverandørnr. TRCB62510250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8
CAS 1189968-99-7
Molekylær formel C18H16D8N2O5
Mængde 50 mg
Synonym N-Boc-N’-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8, 4-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]-1-piperazinecarboxylic Acid 1,1-Dimethylethyl Ester-d8
InChI formel InChI=1S/C18H24N2O5/c1-18(2,3)25-17(22)20-10-8-19(9-11-20)16(21)15-12-23-13-6-4-5-7-14(13)24-15/h4-7,15H,8-12H2,1-3H3/i8D2,9D2,10D2,11D2
SMIL [2H]C1([2H])N(C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])N(C(=O)C2COc3ccccc3O2)C1([2H])[2H]
IUPAC navn tert-butyl 2,2,3,3,5,5,6,6-octadeuterio-4-(2,3-dihydro-1,4-benzodioxine-3-carbonyl)piperazine-1-carboxylate
Molekylvægt (g/mol) 356.44
Formel vægt 356.2187