missing translation for 'onlineSavingsMsg'
Få mere at vide

(4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8, TRC

Artikelnummer. 30334894
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334894 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334894 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28658210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8
CAS 23173-76-4
Molekylær formel C15H14D8N2O2
Mængde 10 mg
Synonym Ethyl 2-(4-Benzylpiperazin-1-yl)acetate, Ethyl 4-Benzyl-1-piperazineacetate, 4-Benzyl-1-piperazineacetic Acid Ethyl Ester, 4-(Phenylmethyl)-1-piperazineacetic Acid Ethyl Ester
InChI formel InChI=1S/C15H22N2O2/c1-2-19-15(18)13-17-10-8-16(9-11-17)12-14-6-4-3-5-7-14/h3-7H,2,8-13H2,1H3
SMIL [2H]C1([2H])N(CC(=O)OCC)C([2H])([2H])C([2H])([2H])N(Cc2ccccc2)C1([2H])[2H]
IUPAC navn ethyl 2-(4-benzylpiperazin-1-yl)acetate
Molekylvægt (g/mol) 270.4
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.