missing translation for 'onlineSavingsMsg'
Få mere at vide

(4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8, TRC

Artikelnummer. 30345717
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345717 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30345717 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2865825MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale (4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8
CAS 23173-76-4
Molekylær formel C15H14D8N2O2
Mængde 5 mg
Synonym Ethyl 2-(4-Benzylpiperazin-1-yl)acetate, Ethyl 4-Benzyl-1-piperazineacetate, 4-Benzyl-1-piperazineacetic Acid Ethyl Ester, 4-(Phenylmethyl)-1-piperazineacetic Acid Ethyl Ester
InChI formel InChI=1S/C15H22N2O2/c1-2-19-15(18)13-17-10-8-16(9-11-17)12-14-6-4-3-5-7-14/h3-7H,2,8-13H2,1H3
SMIL [2H]C1([2H])N(CC(=O)OCC)C([2H])([2H])C([2H])([2H])N(Cc2ccccc2)C1([2H])[2H]
IUPAC navn ethyl 2-(4-benzylpiperazin-1-yl)acetate
Molekylvægt (g/mol) 270.4
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.