Få mere at vide
(4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8, TRC
Artikelnummer. 30335604
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30335604 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335604 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28658250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (4-Benzylpiperazin-1-yl)acetic Acid Ethyl Ester-d8
CAS 23173-76-4
Molekylær formel C15H14D8N2O2
Mængde 50 mg
Synonym Ethyl 2-(4-Benzylpiperazin-1-yl)acetate, Ethyl 4-Benzyl-1-piperazineacetate, 4-Benzyl-1-piperazineacetic Acid Ethyl Ester, 4-(Phenylmethyl)-1-piperazineacetic Acid Ethyl Ester
InChI formel InChI=1S/C15H22N2O2/c1-2-19-15(18)13-17-10-8-16(9-11-17)12-14-6-4-3-5-7-14/h3-7H,2,8-13H2,1H3
SMIL [2H]C1([2H])N(CC(=O)OCC)C([2H])([2H])C([2H])([2H])N(Cc2ccccc2)C1([2H])[2H]
IUPAC navn ethyl 2-(4-benzylpiperazin-1-yl)acetate
Molekylvægt (g/mol) 270.4