missing translation for 'onlineSavingsMsg'
Learn More
4-Benzyloxybenzaldehyde, 98% Product Code.: 11488466

4-Benzyloxybenzaldehyde, 98%

Product Code. p-7022937
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11488466

Brand: Thermo Scientific Alfa Aesar A15934.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4397-53-9 | C14H12O2 | 212.25 g/mol

4-Benzyloxybenzaldehyde is used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand. It is a position isomer of the adenylyl cyclase activator 2-benzyloxybenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyloxybenzaldehyde is used in the synthesis of (5-fluoro-(2R*,3S*)-2,3-bis(4-hydroxyphenyl)pentanenitrile),an estrogen receptor β-selective ligand. It is a position isomer of the adenylyl cyclase activator 2-benzyloxybenzaldehyde.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4397-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00003387,MFCD01632880
InChI Key ZVTWZSXLLMNMQC-UHFFFAOYSA-N
Synonym 4-benzyloxybenzaldehyde, 4-benzyloxy benzaldehyde, benzaldehyde, 4-phenylmethoxy, p-benzyloxy benzaldehyde, benzaldehyde, p-benzyloxy, 4-benzyloxy-benzaldehyde, 4-phenylmethoxy benzaldehyde, 4-benzyl-oxy benzaldehyde, wang aldehyde resin
PubChem CID 78109
IUPAC Name 4-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 69°C to 74°C
Boiling Point 197°C to 199°C (20 mmHg)
Quantity 25 g
Beilstein 1242385
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 4-Benzyloxybenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Benzyloxybenzaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.