Få mere at vide
4-Benzyloxy Atorvastatin-d5 Acetonide tert-Butyl Ester, TRC
Artikelnummer. 30348097
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30348097 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348097 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28765210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Benzyloxy Atorvastatin-d5 Acetonide tert-Butyl Ester
CAS 265989-37-5
Molekylær formel C47H48D5FN2O6
Mængde 10 mg
Synonym (4R,6R)-6-[2-[2-(4-fluorophenyl)-5-(1-methylethyl)-3-(phenyl-d5)-4-[[[4-(phenylmethoxy)phenyl]amino]carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, (6-{2-[3-(4-Benzyloxy-phenylcarbamoyl)-5-(4-fluoro-phenyl)-2-isopropyl-4-phenyl-d5-pyrrol-1-yl]-ethyl}-2,2-dimethyl-[1,3]-dioxane-4-yl)-acetic Acid, tert-Butyl Ester
InChI formel InChI=1S/C47H53FN2O6/c1-31(2)43-42(45(52)49-36-22-24-37(25-23-36)53-30-32-14-10-8-11-15-32)41(33-16-12-9-13-17-33)44(34-18-20-35(48)21-19-34)50(43)27-26-38-28-39(55-47(6,7)54-38)29-40(51)56-46(3,4)5/h8-25,31,38-39H,26-30H2,1-7H3,(H,49,52)/i9D,12D,13D,16D,17D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c(C(=O)Nc3ccc(OCc4ccccc4)cc3)c(C(C)C)n(CCC5CC(CC(=O)OC(C)(C)C)OC(C)(C)O5)c2c6ccc(F)cc6
IUPAC navn tert-butyl 2-[6-[2-[2-(4-fluorophenyl)-3-(2,3,4,5,6-pentadeuteriophenyl)-4-[(4-phenylmethoxyphenyl)carbamoyl]-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molekylvægt (g/mol) 765.96
Formel vægt 765.4201